BAF378631000
Scienceware® colony counter system
Complete System
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
General description
This small, efficient colony counting system with magnifier and counting grid is CE approved and available complete or each item may be purchased separately. The complete system includes:
Replacement pens and battery for the colony counter are available.
- Pen-style colony counter
- Mini-magnifier
- Mini light box II
- AC line current adapter
- Counting grid
Replacement pens and battery for the colony counter are available.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)