Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H18BrClN2O6
CAS Number:
Molecular Weight:
449.68
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
assay
≥95%
form
solid
storage temp.
−20°C
SMILES string
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2c[nH]c3ccc(Br)c(Cl)c23
InChI
1S/C16H18BrClN2O6/c1-6(22)20-13-15(24)14(23)10(5-21)26-16(13)25-9-4-19-8-3-2-7(17)12(18)11(8)9/h2-4,10,13-16,19,21,23-24H,5H2,1H3,(H,20,22)/t10-,13-,14-,15-,16-/m1/s1
InChI key
SUWPNTKTZYIFQT-XZINFULNSA-N
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.