Skip to Content
Merck

B9012

3-Bromopropylamine hydrobromide

Synonym(s):

3-Aminopropyl bromide hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
BrCH2CH2CH2NH2 · HBr
CAS Number:
Molecular Weight:
218.92
EC Number:
225-675-0
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
3906418
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

171-172 °C (lit.)

SMILES string

Br.NCCCBr

InChI

1S/C3H8BrN.BrH/c4-2-1-3-5;/h1-3,5H2;1H

InChI key

PQIYSSSTRHVOBW-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library