Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C17H23NO6
CAS Number:
Molecular Weight:
337.37
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Beilstein/REAXYS Number:
2482076
Product Name
Boc-Glu-OBzl,
assay
≥98% (TLC)
form
powder
color
white to off-white
mp
95-99 °C
application(s)
peptide synthesis
storage temp.
−20°C
SMILES string
CC(C)(C)OC(=O)N[C@@H](CCC(O)=O)C(=O)OCc1ccccc1
InChI
1S/C17H23NO6/c1-17(2,3)24-16(22)18-13(9-10-14(19)20)15(21)23-11-12-7-5-4-6-8-12/h4-8,13H,9-11H2,1-3H3,(H,18,22)(H,19,20)/t13-/m0/s1
InChI key
CVZUKWBYQQYBTF-ZDUSSCGKSA-N
General description
Substrate for Vitamin K-dependent carboxylation.
Biochem/physiol Actions
Boc-Glu-OBzl is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate benzyl ester residues.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.