Skip to Content
Merck

B7775

[Hyp3]-Bradykinin

≥97% (HPLC), suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H73N15O12
CAS Number:
Molecular Weight:
1076.21
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

[Hyp3]-Bradykinin, ≥97% (HPLC)

assay

≥97% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

N[C@@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)N2C[C@H](O)C[C@H]2C(=O)NCC(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CO)C(=O)N4CCC[C@H]4C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C50H73N15O12/c51-32(15-7-19-56-49(52)53)45(73)64-22-10-18-38(64)47(75)65-27-31(67)25-39(65)43(71)58-26-40(68)59-34(23-29-11-3-1-4-12-29)41(69)62-36(28-66)46(74)63-21-9-17-37(63)44(72)61-35(24-30-13-5-2-6-14-30)42(70)60-33(48(76)77)16-8-20-57-50(54)55/h1-6,11-14,31-39,66-67H,7-10,15-28,51H2,(H,58,71)(H,59,68)(H,60,70)(H,61,72)(H,62,69)(H,76,77)(H4,52,53,56)(H4,54,55,57)/t31-,32+,33+,34+,35+,36+,37+,38+,39+/m1/s1

InChI key

JXRLHZCEMXTCBN-DIBGMJQNSA-N

Gene Information

rat ... Tac1(24806)

Biochem/physiol Actions

[Hyp3]-Bradykinin is a bradykinin receptor agonist used to differentiate bradykinin receptors in various cells, tissues and species.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library