Skip to Content

B7541

(2R,3R,4E)-1,3-O-Benzylidene-4-octadecene-1,2,3-triol

≥98%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H40O3
CAS Number:
Molecular Weight:
388.58
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98%

SMILES string

CCCCCCCCCCCCC\C=C\[C@H]1O[C@H](OC[C@H]1O)c2ccccc2

Other Notes

Protected alcohol analog of D-sphingosine.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library