Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
EC Number:
205-883-8
UNSPSC Code:
12352103
PubChem Substance ID:
Beilstein/REAXYS Number:
1913029
MDL number:
bp
>500 °C (lit.)
mp
277-279 °C (lit.)
SMILES string
c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56
InChI
1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H
InChI key
GYFAGKUZYNFMBN-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Benzo[ghi]perylene
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Souleymane Coulibaly et al.
Biological & pharmaceutical bulletin, 38(9), 1395-1403 (2015-09-04)
To clarify the seasonal fluctuations in air pollution and the effect of long-range transport, we collected airborne particles (n=118) at Dazaifu in Fukuoka, Japan, from June 2012 to May 2013 and measured Pb and SO4(2-), which are indicators of the