Skip to Content
Merck

B5765

O-tert-Butylhydroxylamine hydrochloride

≥99%

Synonym(s):

2-Aminooxy-2-methylpropane hydrochloride, O-(1,1-Dimethylethyl)hydroxylamine hydrochloride, tert-Butoxyamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)3CONH2 · HCl
CAS Number:
Molecular Weight:
125.60
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
254-590-1
Beilstein/REAXYS Number:
3668106
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥99%

form

powder

SMILES string

Cl.CC(C)(C)ON

InChI

1S/C4H11NO.ClH/c1-4(2,3)6-5;/h5H2,1-3H3;1H

InChI key

ZBDXGNXNXXPKJI-UHFFFAOYSA-N

Application

Reactant involved in synthesis of biologically active molecules including:
  • CGS 25966 derivatives for use as MMP inhibitors
  • Imidazolidinedione derivatives for use as antimalarial treatments
  • Pyrimidine ribonucleotide analogs as P2Y6 receptor agonists
  • Rab proteins for isoprenylation and geranylgeranylation inhibition

Reactant involved in:
  • Synthesis of N-(arylethyl)-O-tert-butylhydroxamates for use as Weinreb amide equivalents
  • Double allylic alkylation of indole-2-hydroxamates
  • SN2 substitution reactions at amide nitrogens
  • Photocycloaddition to C=N bonds for synthesis of 1,3-diazepines

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service