Skip to Content

B5250

6-Benzoyl-2-naphthyl phosphate disodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H11O5PNa2
CAS Number:
Molecular Weight:
372.22
UNSPSC Code:
12352100
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

[Na+].[Na+].[O-]P([O-])(=O)Oc1ccc2cc(ccc2c1)C(=O)c3ccccc3





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library