Skip to Content
Merck

B4258

Bay oil

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


refractive index

n20/D 1.513 (lit.)

bp

245 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

lipid type

oils



pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

134.6 °F - closed cup

flash_point_c

57 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library