Skip to Content
Merck
All Photos(1)

Key Documents

B4152

Sigma-Aldrich

4-Biphenylacetic acid

≥98% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2CO2H
CAS Number:
Molecular Weight:
212.24
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Assay

≥98% (titration)

mp

159-160 °C (lit.)

SMILES string

OC(=O)Cc1ccc(cc1)-c2ccccc2

InChI

1S/C14H12O2/c15-14(16)10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H,10H2,(H,15,16)

InChI key

QRZAKQDHEVVFRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Yi Mok et al.
Journal of medicinal chemistry, 56(5), 1843-1852 (2013-02-05)
β-Secretase (BACE1), the enzyme responsible for the first and rate-limiting step in the production of amyloid-β peptides, is an attractive target for the treatment of Alzheimer's disease. In this study, we report the application of the de novo fragment-based molecular
R F Halliwell et al.
Neuropharmacology, 43(4), 778-787 (2002-10-09)
A hybrid molecule composed of the antimicrobial, norfloxacin, linked to the non-steroidal anti-inflammatory drug (NSAID), biphenylacetic acid, which we have termed norbiphen, is a lethal convulsant in vivo and an antagonist of rodent GABA(A) receptors in vitro. In the present
Miho Sekiguchi et al.
BMC musculoskeletal disorders, 9, 15-15 (2008-02-01)
External medication that is absorbed percutaneously may be used to reduce inflammation and relieve pain from acute injuries such as ankle sprains and bruises. The plaster method of percutaneous absorption for non-steroidal anti-inflammatory drugs (NSAIDs) was established in Japan in
Liqing Zang et al.
Zebrafish, 8(4), 203-210 (2011-12-21)
A novel protocol using gluten as a carrier material was developed to administer chemicals to adult zebrafish, per os (p.o.). To evaluate the capacity of gluten to retain chemicals, we prepared gluten granules containing eight types of chemicals with different
Ananda Sarkar et al.
Journal of molecular modeling, 15(10), 1221-1228 (2009-03-07)
Cyclooxygenase (COX) is a well-known enzyme, which converts arachidonic acid to prostaglandins H(2) (PGH(2)), which are the effective mediators of inflammation. 4', 5 di-substituted 3-biphenyl acetic acids (BPA) and several alpha-methyl derivatives (MBPA) of it are widely used as powerful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service