Skip to Content
Merck
All Photos(1)

Key Documents

B2395

Sigma-Aldrich

4-Bromoaniline

~98%, crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NH2
CAS Number:
Molecular Weight:
172.02
Beilstein:
742031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

~98%

form

crystalline

color

white to light yellow

mp

56-62 °C (lit.)

SMILES string

Nc1ccc(Br)cc1

InChI

1S/C6H6BrN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

WDFQBORIUYODSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Major et al.
Xenobiotica; the fate of foreign compounds in biological systems, 33(8), 855-869 (2003-08-26)
1. The metabolic fate of 4-bromoaniline (4-BrA) was investigated in rat following intraperitoneal administration at 50 mg kg(-1) using HPLC-TOF-MS/MS. 2. The sensitivity provided by the use of TOF-MS/MS, aided by the distinctive isotope pattern resulting from the presence of
Spectrophotometric determination of carnosine, anserine, and taurine in skeletal muscle.
C J Parker
Analytical biochemistry, 108(2), 303-305 (1980-11-01)
G B Scarfe et al.
Xenobiotica; the fate of foreign compounds in biological systems, 32(4), 325-337 (2002-05-25)
1. The urinary excretion of 4-bromoaniline and its [carbonyl-(13)C]-labelled N-acetanilide, together with their corresponding metabolites, have been investigated in the rat following i.p. administration at 50 mg kg(-1). 2. Metabolite profiling was performed by reversed-phase HPLC with UV detection, whilst
F Flores-Céspedes et al.
Environmental pollution (Barking, Essex : 1987), 142(3), 449-456 (2006-01-03)
In this study we have evaluated the effects of dissolved organic matter (DOM) on sorption of imidacloprid, 3,4-dichloroaniline (3,4-DCA) and 4-bromoaniline (4-BA) on a typical calcareous soil (Luvic Xerosol) from south-eastern Spain. Two different types of DOM were used, that
M C Roberts et al.
Equine veterinary journal, 11(4), 239-243 (1979-10-01)
The absorption of d-xylose forms the basis of a useful screening test in the investigation of small intestinal disorders in the horse. A comparison has been made of different assay methods and there was no significant difference between the results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service