Skip to Content
Merck
All Photos(1)

Key Documents

B1936

Sigma-Aldrich

Borrelidin

from Streptomyces sp., ≥95% (HPLC), lyophilized powder

Synonym(s):

2-(7-Cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxooxacyclooctadeca-4,6-dien-2-yl)-cyclopentanecarboxylic acid, Borrelidine, Cyclopentanecarboxylic acid, NSC 216128, Treponemycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H43NO6
CAS Number:
Molecular Weight:
489.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Streptomyces sp.

Assay

≥95% (HPLC)

form

lyophilized powder

solubility

DMF: soluble
DMSO: soluble
ethanol: soluble
ethyl acetate: soluble
methanol: soluble

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

[H][C@]1(CCC[C@H]1C(O)=O)[C@@H]2C\C=C\C=C(C#N)[C@H](O)[C@@H](C)C[C@H](C)C[C@H](C)C[C@H](C)[C@@H](O)CC(=O)O2

InChI

1S/C28H43NO6/c1-17-12-18(2)14-20(4)27(32)21(16-29)8-5-6-11-25(22-9-7-10-23(22)28(33)34)35-26(31)15-24(30)19(3)13-17/h5-6,8,17-20,22-25,27,30,32H,7,9-15H2,1-4H3,(H,33,34)/b6-5+,21-8-/t17-,18+,19-,20-,22+,23+,24-,25-,27+/m0/s1

InChI key

OJCKRNPLOZHAOU-UGKRXNSESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Biochem/physiol Actions

Borrelidin is a potent angiogenesis inhibitor that induces apoptosis in capillary tube-forming cells. Also displays antimalarial activity against drug-resistant Plasmodia. Antimicrobial and selective threonyl t-RNA synthetase inhibitor.

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kira J Weissman
Trends in biotechnology, 25(4), 139-142 (2007-02-20)
Mutasynthesis couples the power of chemical synthesis with molecular biology to generate derivatives of medicinally valuable, natural products. Recently, this technique has been exploited by Cambridge-based biotech company Biotica Technology Ltd, and their collaborators, to generate promising new variants of
Steven J Moss et al.
Chemical communications (Cambridge, England), (22)(22), 2341-2343 (2006-05-31)
We report the directed biosynthesis of borrelidin analogues and their selective anti-proliferative activity against human cancer cell lines.
Chongxi Liu et al.
International journal of systematic and evolutionary microbiology, 63(Pt 3), 1030-1036 (2012-06-19)
A borrelidin-producing actinomycete, designated strain NEAU-W2(T), was isolated from the root surface of soybean [Glycine max (L.) Merr] and characterized using a polyphasic approach. The organism was found to have morphological and chemotaxonomic characteristics typical of streptomycetes. The G+C content
Carlos Olano et al.
Molecular microbiology, 52(6), 1745-1756 (2004-06-10)
The 18-membered polyketide macrolide borrelidin exhibits a number of important biological activities, including potent angiogenesis inhibition. This has prompted two recent total syntheses as well as the cloning of the biosynthetic gene cluster from Streptomyces parvulus Tü4055. Borrelidin possesses some
Barrie Wilkinson et al.
Bioorganic & medicinal chemistry letters, 16(22), 5814-5817 (2006-09-12)
A set of novel borrelidin analogues have been prepared by precursor-directed biosynthesis. Structure-activity relationship analysis suggests that steric structural arrangement within the C17 side chain is important for differentiating cytotoxic and anti-angiogenic activities. A C17-cyclobutyl analogue 3 was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service