Skip to Content
Merck

B1136

Boc-Val-Leu-Lys-7-amido-4-methylcoumarin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H49N5O7
CAS Number:
Molecular Weight:
615.76
UNSPSC Code:
12352204
PubChem Substance ID:
EC Number:
277-536-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(NC(=O)OC(C)(C)C)C(C)C)C(=O)NC(CCCCN)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library