Skip to Content
Merck

B1061

BN82002 hydrochloride

≥97% (HPLC), solid

Synonym(s):

4-Dimethylamino-2-methoxy-6-((methyl-[2-(4-nitrophenyl)ethyl]amino)methyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H25N3O4 · xHCl
CAS Number:
Molecular Weight:
359.42 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

form

solid

color

tan

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].COc1cc(cc(CN(C)CCc2ccc(cc2)[N+]([O-])=O)c1O)N(C)C

Biochem/physiol Actions

BN82002 is a potent, selective, and irreversible CDC25 phosphatase family inhibitor.
BN82002 is a potent, selective, and irreversible CDC25 phosphatase family inhibitor. BN82002 displays ~20-fold greater selectivity over CD45 tyrosine phosphatase (IC50 in μM = 2.4, 3.9, 6.3, 5.4, and 4.6 for 25A, 25B2, 25B3, 25C, and 25C-cat, respectively). BN82002 has been shown to delay cell cycle progression in vitro (IC50 in the range of 7.2 - 32.6 μM). BN82002 reduces tumor growth in athymic mice xenografted with the human pancreatic cell MIA PaCa-2 (15 mg/kg, i.p. route).

Disclaimer

Hygroscopic.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library