Skip to Content
Merck

B0755

o-Bromoaniline

Synonym(s):

2-Bromoaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
210-421-3
UNSPSC Code:
12352103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C



signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library