Skip to Content
Merck
All Photos(1)

Key Documents

AXYTR222CSTKS

Pipette reference tips

universal fit, stacked rack, size 200 μL, pkg of 5x960tips/cs, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
size:
200 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TR-222-C-STK-S
Corning

sterility

sterile

manufacturer/tradename

Axygen Inc TR-222-C-STK-S
Corning

packaging

pkg of 5x960tips/cs

size

200 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service