Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF200

Pipette filter tips

universal fit, size 200 μL, pkg of 10x1000tips/cs

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CZK 28,500.00

CZK 28,500.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
CZK 28,500.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
manufacturer/tradename:
Axygen Inc TF-200
Corning
size:
200 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier
endotoxin free
filter
packaging:
pkg of 10x1000tips/cs

CZK 28,500.00


Please contact Customer Service for Availability

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-200
Corning

packaging

pkg of 10x1000tips/cs

size

200 μL

Looking for similar products? Visit Product Comparison Guide

Application

Axygen® Filter Barrier tips are suitable for DNA amplification and microbiological applications where slight contamination can alter the results.

Features and Benefits

Axygen® Filter Barrier tips have the following features and benefits:
  • Tips create a physical barrier to prevent cross-contamination and block aerosols
  • The DNA inhibitors free-polyethylene filters are fit into the tips robotically to avoid nuclease contamination
  • Packed in sealed-bottom racks
  • Tips are RNAse and DNAse free
  • Non-pyrogenic

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service