Skip to Content
Merck

Skip To

AXYSCOR

Screw caps with O-rings

red, pkg of 8x500caps/cs

Synonym(s):

Corning screw cap

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 pkg
Please contact Customer Service for Availability
25 600,00 CZK

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Color:
red
Packaging:
pkg of 8x500caps/cs

25 600,00 CZK


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Axygen Inc SCO-R, Corning

packaging

pkg of 8x500caps/cs

color

red

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AXYSCOBRAXYSCOGAXYSCOY
description

red, pkg of 8x500caps/cs

description

brown, pkg of 8x500caps/cs

description

green, pkg of 8x500caps/cs

description

yellow, pkg of 8x500caps/cs

color

red

color

brown

color

green

color

yellow

manufacturer/tradename

Axygen Inc SCO-R, Corning

manufacturer/tradename

Axygen Inc SCO-BR, Corning

manufacturer/tradename

Axygen Inc SCO-G, Corning

manufacturer/tradename

Axygen Inc SCO-Y, Corning

packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs

packaging

pkg of 8x500caps/cs


Still not finding the right product?

Explore all of our products under Screw caps with O-rings




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library