Skip to Content
Merck
All Photos(2)

Key Documents

AXYPCR96FSCS

Corning® Axygen® 96 Well PCR Amplification Plate

full skirt, 96 well, barcoded, clear, sterile

Synonym(s):

PCR amplification plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
material:
clear
clear polypropylene wells
size:
96 wells
Product AXYPCR96FSCS is not currently sold in your country. Contact Technical Service

material

clear
clear polypropylene wells

sterility

sterile

feature

skirt

packaging

case of 50 ea

manufacturer/tradename

Axygen Inc
Corning

technique(s)

PCR: suitable

size

96 wells

working volume

200 mL

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service