Skip to Content
Merck
All Photos(1)

Key Documents

AXYMCT060C

Microtubes

size 0.6 mL, clear, pkg of 10x1000tubes/cs

Synonym(s):

Corning tubes, cap tubes, polypropylene tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
CZK 14,800.00

CZK 14,800.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
CZK 14,800.00

About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

CZK 14,800.00


Please contact Customer Service for Availability

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

non-sterile

feature

RNase and DNase free
endotoxin free
graduations

manufacturer/tradename

Axygen Inc MCT-060-C
Corning

packaging

pkg of 10x1000tubes/cs

size

0.6 mL

color

clear
clear

fitting

cap (push fit)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide

General description

  • Axygen® MaxyClear homopolymer microtubes offer high tensile strength and chemical resistance.
  • Available with a snap cap that ensures an efficient seal for sample protection.
  • Frosted surface for labeling.
  • A piercing port in the center of the cap with a thin membrane allows easy access to samples by syringes.
  • Free of RNase/DNase and nonpyrogenic.
  • Maximum G-force is 14,000 RCF.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service