Skip to Content
Merck
All Photos(1)

Key Documents

AXYBF444

Breathable sealing film for tissue culture plates, deep well plates

96 well, for cell growth

Synonym(s):

plate sealing mat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
Pricing and availability is not currently available.

form

film

manufacturer/tradename

Axygen Inc
Corning


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service