Skip to Content
Merck
All Photos(1)

Key Documents

AV51270

Sigma-Aldrich

Anti-TNNI2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-AMCD2B, Anti-DA2B, Anti-FSSV, Anti-Troponin I type 2 (skeletal, fast)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 7,440.00

CZK 7,440.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 7,440.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 7,440.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

21 kDa

species reactivity

bovine, dog, horse, guinea pig, human, rat, rabbit, goat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNNI2(7136)

Immunogen

Synthetic peptide directed towards the N terminal region of human TNNI2

Application

Anti-TNNI2 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml.

Biochem/physiol Actions

Troponin I type 2 (TNNI2) is a fast-twitch muscle protein belonging to the troponin I gene family. It forms a complex that regulates striated muscle contraction along with troponins T and C. TNNI2 also has a role in smooth muscle contraction, angiogenesis, metastasis and tumor growth. Mutations in TNNI2 gene have been implicated in myopathy1 and distal arthrogryposis type 2B.

Sequence

Synthetic peptide located within the following region: PLHIPGSMSEVQELCKQLHAKIDAAEEEKYDMEVRVQKTSKELEDMNQKL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miao Jiang et al.
Human genetics, 120(2), 238-242 (2006-06-28)
Distal arthrogryposis (DA) is composed of a group of clinically and genetically heterogeneous disorders, characterized by multiple congenital contractures of the limbs. Point mutations in three genes encoding contractile fast-twitch myofibers, TPM2, TNNI2 and TNNT3, were recently identified in DA
GuangWu Xiong et al.
Science in China. Series C, Life sciences, 50(1), 93-100 (2007-03-30)
To explore the efficiency and mechanism of ovarian carcinoma gene therapy with the human fast-twitch skeletal muscle troponin I gene (Tnl-fast), Tnl-fast cDNA was transferred into human ovarian adenocarcinoma cell-line SK-OV-3. In vitro, the cell growth and cell cycle of
E Kimber et al.
Neurology, 67(4), 597-601 (2006-08-23)
To describe a three-generation family with distal arthrogryposis associated with myopathy and caused by a mutation in the gene encoding for sarcomeric thin filament protein troponin I, TNNI2. The authors performed clinical investigations and reviewed medical records. Muscle biopsy specimens

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service