Skip to Content
Merck
All Photos(1)

Key Documents

AV50276

Sigma-Aldrich

Anti-ASB5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Ankyrin repeat and SOCS box-containing 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,600.00

CZK 9,600.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 9,600.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,600.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

rabbit, mouse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASB5(140458)

Immunogen

Synthetic peptide directed towards the C terminal region of human ASB5

Application

Anti-ASB5 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Ankyrin repeat and SOCS box containing 5 (ASB5) belongs to the ankyrin repeat and SOCS box-containing (ASB) family of proteins. These proteins regulate protein turnover by targeting proteins for polyubiquitination and proteasome-mediated degradation. It is expressed in endothelial cells and smooth muscle cells and is involved in arteriogenesis.

Sequence

Synthetic peptide located within the following region: LLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin T Kile et al.
Trends in biochemical sciences, 27(5), 235-241 (2002-06-22)
Although initially identified in the suppressor of cytokine signaling (SOCS) family of proteins, the C-terminal SOCS box has now been identified in more than 40 proteins in nine different families. Growing evidence suggests that the SOCS box, similar to the
Kerstin Boengler et al.
Biochemical and biophysical research communications, 302(1), 17-22 (2003-02-21)
Arteriogenesis, the growth of pre-existing collateral arteries, can be induced in rabbit by occlusion of the femoral artery. In order to identify and characterize genes differentially expressed during the early phase of arteriogenesis, cDNA of collateral arteries 24h after femoral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service