Skip to Content
Merck

Skip To

AV48436

Anti-HSPB6 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ32389, Anti-Heat shock protein, α-crystallin-related, B6, Anti-Hsp20

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPB6(126393)

General description

HSPB6 (Hsp20) encodes a heat shock protein that may be involved in the relaxation of smooth muscles. It is known to interact with Bag3. HSPB6 has also been implicated in platelet aggregation, atherosclerosis, myocardial infarction, insulin resistance and Alzheimer′s disease.
Rabbit Anti-HSPB6 antibody recognizes canine, bovine, human, mouse, rat, chicken, and pig HSPB6.

Immunogen

Synthetic peptide directed towards the middle region of human HSPB6

Application

Rabbit Anti-HSPB6 antibody is suitable for western blot applications at a concentration of 1.25μg/ml.

Biochem/physiol Actions

HSPB6 is associated with actin and modulates smooth muscle relaxation.HSPB6 is associated with actin (see MIM 102540) and modulates smooth muscle relaxation (Tessier et al., 2003 [PubMed 12842460]).[supplied by OMIM].

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: ARHEERPDEHGFVAREFHRRYRLPPGVDPAAVTSALSPEGVLSIQAAPAS

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters