Skip to Content
Merck
All Photos(3)

Key Documents

AV42290

Sigma-Aldrich

Anti-AGR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AG2, Anti-Anterior gradient homolog 2 (Xenopus laevis), Anti-GOB-4, Anti-HAG-2, Anti-XAG-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 12,800.00

CZK 12,800.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 12,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 12,800.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

20 kDa

species reactivity

rat, dog, rabbit, human, bovine, horse, mouse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGR2(10551)

General description

Anterior gradient homolog 2 (Xenopus laevis) (AGR2) is a protein disulfide isomerase (PDI) which aids protein folding and assembly by catalyzing formation and shuffling of cysteine disulfide bonds in the endoplasmic reticulum (ER). AGR2 is present in the ER of intestinal secretory epithelial cells and it is believed to have a role in mucin processing and colitis.

Specificity

Anti-AGR2 antibody polyclonal antibody reacts with the bovine, chicken, zebrafish, human, mouse, and rat anterior gradient homolog 2 (Xenopus laevis).

Immunogen

Synthetic peptide directed towards the middle region of human AGR2

Application

Anti-anterior gradient homolog 2 (Xenopus laevis) polyclonal antibody is used to tag anterior gradient homolog 2 (Xenopus laevis) protein(s)/subunits for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques.

Biochem/physiol Actions

AGR2 and hAG-3, human homologues of genes involved in differentiation, are associated with oestrogen receptor-positive breast tumours and interact with metastasis gene C4.4a and dystroglycan. Increased AGR2 expression is a valuable prognostic factor to predict the clinical outcome of the prostate cancer patients.

Sequence

Synthetic peptide located within the following region: AEQFVLLNLVYETTDKHLSPDGQYVPRIMFVDPSLTVRADITGRYSNRLY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service