Skip to Content
Merck

Skip To

AV41594

Anti-C11ORF54 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PTD012

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

rat, mouse, horse, rabbit, guinea pig, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C11orf54(28970)

General description

PTD012 (C11ORF54) is a zinc-binding nuclear protein that exhibits ester hydrolase activity using the substrate p-nitrophenyl acetate. The biological role of C11ORF54 has not been determined.

Immunogen

Synthetic peptide directed towards the C terminal region of human C11orf54

Application

Anti-C11ORF54 (AB2) polyclonal antibody is used to tag C11ORF54 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the distribution and role of PTD012.

Biochem/physiol Actions

Anti-C11ORF54 (AB2) polyclonal antibody reacts with, bovine, human, mouse, rat, and canine PTD012 proteins.
C11orf54 exhibits ester hydrolase activity on the substrate p-nitrophenyl acetate.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: PVFVSRDPGFDLRLEHTHFFSRHGEGGHYHYDTTPDIVEYLGYFLPAEFL

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters