Skip to Content
Merck
All Photos(3)

Key Documents

AV32618

Sigma-Aldrich

Anti-HDAC2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Histone deacetylase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 8,700.00

CZK 8,700.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1208


Select a Size

Change View
100 μL
CZK 8,700.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CZK 8,700.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1208

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

rat, mouse, rabbit, dog, pig, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC2(3066)

General description

HDAC2 is a member of the deacetylase family that forms transcriptional repressor complexes. Studies have reported that HDAC2 negatively regulates synaptic plasticity, learning and memory functions. In colorectal carcinogenesis, loss of APC is known to induce HDAC2 expression.
Rabbit Anti-HDAC2 recognizes bovine, human, canine, chicken, rat, and mouse HDAC2.

Immunogen

Synthetic peptide directed towards the C terminal region of human HDAC2

Application

Rabbit Anti-HDAC2 antibody can be used for western blot (0.2-2.0μg/ml) and IHC (4-8μg/ml) applications.

Biochem/physiol Actions

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly concerved from yeast to human.

Sequence

Synthetic peptide located within the following region: EEFSDSEDEGEGGRRNVADHKKGAKKARIEEDKKETEDKKTDVKEEDKSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Zhu et al.
Cancer cell, 5(5), 455-463 (2004-05-18)
Inappropriate transcriptional repression involving histone deacetylases (HDACs) is a prominent cause for the development of leukemia. We now identify faulty expression of a specific mediator of transcriptional repression in a solid tumor. Loss of the adenomatosis polyposis coli (APC) tumor
Ji-Song Guan et al.
Nature, 459(7243), 55-60 (2009-05-09)
Chromatin modifications, especially histone-tail acetylation, have been implicated in memory formation. Increased histone-tail acetylation induced by inhibitors of histone deacetylases (HDACis) facilitates learning and memory in wild-type mice as well as in mouse models of neurodegeneration. Harnessing the therapeutic potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service