Skip to Content
Merck
All Photos(1)

Key Documents

AV32524

Sigma-Aldrich

Anti-NR5A2 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-B1F, Anti-B1F2, Anti-CPF, Anti-FTF, Anti-FTZ-F1, Anti-FTZ-F1beta, Anti-LRH-1, Anti-Nuclear receptor subfamily 5, group A, member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,600.00

CZK 9,600.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 9,600.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,600.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

guinea pig, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR5A2(2494)

General description

NR5A2 belongs to the nuclear receptor subfamily. It has been implicated in acinar cell differentiation and pancreatic carcinogenesis.
Rabbit Anti-NR5A2 (AB2) antibody recognizes chicken, human, mouse, and rat NR5A2.

Immunogen

Synthetic peptide directed towards the middle region of human NR5A2

Application

Rabbit Anti-NR5A2 (AB2) antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

NR5A2 binds to the sequence element 5′-AACGACCGACCTTGAG-3′ of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. It may be responsable for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. It is a key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. It may also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.

Sequence

Synthetic peptide located within the following region: LPPTDYDRSPFVTSPISMTMLHGSLQGYQTYGHFPSRAIKSEYPDPYTSS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Flandez et al.
Gut, 63(4), 647-655 (2013-04-20)
Nr5a2 participates in biliary acid metabolism and is a major regulator of the pancreatic exocrine programme. Single nucleotide polymorphisms in the vicinity of NR5A2 are associated with the risk of pancreatic ductal adenocarcinoma (PDAC). To determine the role of Nr5a2
Guido von Figura et al.
Gut, 63(4), 656-664 (2013-05-07)
Emerging evidence from mouse models suggests that mutant Kras can drive the development of pancreatic ductal adenocarcinoma (PDA) precursors from acinar cells by enforcing ductal de-differentiation at the expense of acinar identity. Recently, human genome-wide association studies have identified NR5A2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service