Skip to Content
Merck
All Photos(3)

Key Documents

AV31433

Sigma-Aldrich

Anti-POU6F1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-POU domain, class 6, transcription factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,600.00

CZK 9,600.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CZK 9,600.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 9,600.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

rabbit, bovine, mouse, horse, rat, human, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POU6F1(5463)

General description

POU6F1 is a POU homeobox containing transcription factor that represses Oct2A-mediated stimulation. POU6F1 has been implicated in cell proliferation and ovarian adenocarcinoma.
Rabbit Anti-POU6F1 (AB1) recognizes human, mouse, rat, canine, zebrafish, and bovine POU6F1

Immunogen

Synthetic peptide directed towards the middle region of human POU6F1

Application

Rabbit Anti-POU6F1 (AB1) antibody can be used for immunohistochemistry (4-8μg/ml) and western blot (0.12μg/ml) applications.

Biochem/physiol Actions

The POU6F1 gene encodes a protein that is part of a family of transcription factors which exhibit distinct temporal and spatial patterns of expression.

Sequence

Synthetic peptide located within the following region: YFEKNPLPTGQEITEIAKELNYDREVVRVWFCNRRQTLKNTSKLNVFQIP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Wey et al.
Biochemical and biophysical research communications, 220(2), 274-279 (1996-03-18)
Transcription factors of the POU family recognize DNA through their POU domain which represents a bipartite DNA binding motif consisting of a POU specific domain and a POU homeobox. It is thought that both subdomains make specific contacts with DNA
Norihito Yoshioka et al.
Human cell, 22(4), 94-100 (2009-10-31)
Clear cell adenocarcinoma of the ovary often shows resistance to anticancer agents. We investigated new molecules to use when developing molecular-targeting therapy for clear cell adenocarcinoma of the ovary. RMG-I cells without invasive potential and RMG-V cells with invasive potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service