Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
monoclonal
Technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:60
Application:
IHC (p)
Citations:
-
biological source
rabbit
Quality Segment
antibody form
Ig fraction of antiserum
clone
monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:60
shipped in
wet ice
storage temp.
2-8°C
visualization
nuclear
General description
Hepatocyte nuclear factor-4α (HNF4α) is a member of the nuclear receptor superfamily is critical for the maintenance of epithelial cell function and normal colon physiology. Isoforms of HNF4α are generated by alternative splicing of the P1 and P2 promoter. Isoforms of HNF4α 1-6 originate from P1 promoter while HNF4α 7-12 are derived from the P2 promoter. The P1 and P2 proteins differ in approximately 16-29 amino acids in the N-terminal domain. HNF4α is critical for the balance between proliferation and differentiation. In normal tissues, HNF4α P1 is localized to the nucleus and expressed in the small intestine, colon, liver, and kidney, but no expression was found in stomach and lung. HNF4α P1 may be used to identify metastatic tumors to the lung. In gastric carcinomas, intestinal, mixed and null types showed HNF4α P1 positive, but the gastric type was HNF4α P1 negative. In contrast, HNF4α P2 was expressed in all gastric carcinomas. HNF4α P1 antibody may be useful for subclassification of gastric tumors. In several studies focusing on colorectal carcinoma, HNF4α P1 was shown to be frequently lost in tumor cells. In contrast, HNF4α P2 expression was unaffected.
Physical form
Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
For a copy of the IFU and CofA contact IVDorder@ABCAM.com
For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
Still not finding the right product?
Explore all of our products under HNF4 alpha P1 (EP366) Rabbit Monoclonal Primary Antibody
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



