Skip to Content
Merck

A9889

Adipic acid monomethyl ester

Synonym(s):

mono-Methyl adipate, Adipic acid monomethyl ester, Monomethyl adipate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HO2C(CH2)4CO2CH3
CAS Number:
Molecular Weight:
160.17
EC Number:
211-019-0
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
1766411
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


refractive index

n20/D 1.441 (lit.)

bp

162 °C/10 mmHg (lit.)

mp

7-9 °C (lit.)

density

1.08 g/mL at 25 °C (lit.), 1.081 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)CCCCC(O)=O

InChI

1S/C7H12O4/c1-11-7(10)5-3-2-4-6(8)9/h2-5H2,1H3,(H,8,9)

InChI key

UOBSVARXACCLLH-UHFFFAOYSA-N



Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library