Skip to Content
Merck
All Photos(1)

Documents

A9432

Millipore

D-(−)-Arabinose

suitable for component for culture media

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723079
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mp

162-164 °C (lit.)

suitability

suitable for component for culture media

SMILES string

O[C@@H]1COC(O)[C@@H](O)[C@@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3-,4+,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-ZRMNMSDTSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.

Quality

Screened for heavy metal contamination.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuhiro Yamagata et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(2), 578-583 (2014-12-31)
Drosophila melanogaster can acquire a stable appetitive olfactory memory when the presentation of a sugar reward and an odor are paired. However, the neuronal mechanisms by which a single training induces long-term memory are poorly understood. Here we show that
W Joost Wiersinga et al.
Emerging infectious diseases, 21(1), 40-47 (2014-12-23)
Burkholderia pseudomallei, an environmental gram-negative bacillus, is the causative agent of melioidosis and a bio-threat agent. Reports of B. pseudomallei isolation from soil and animals in East and West Africa suggest that melioidosis might be more widely distributed than previously
Maurizio Bettiga et al.
Biotechnology for biofuels, 1(1), 16-16 (2008-10-25)
Ethanolic fermentation of lignocellulosic biomass is a sustainable option for the production of bioethanol. This process would greatly benefit from recombinant Saccharomyces cerevisiae strains also able to ferment, besides the hexose sugar fraction, the pentose sugars, arabinose and xylose. Different
Luke L Lairson et al.
Nature chemical biology, 2(12), 724-728 (2006-10-24)
Despite their unparalleled catalytic prowess and environmental compatibility, enzymes have yet to see widespread application in synthetic chemistry. This lack of application and the resulting underuse of their enormous potential stems not only from a wariness about aqueous biological catalysis
Arie Gruzman et al.
Journal of medicinal chemistry, 51(24), 8096-8108 (2008-12-04)
Type 2 diabetes mellitus has reached epidemic proportions; therefore, the search for novel antihyperglycemic drugs is intense. We have discovered that D-xylose increases the rate of glucose transport in a non-insulin-dependent manner in rat and human myotubes in vitro. Due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service