Skip to Content

A7897

5′-Amino-5′-deoxyadenosine p-toluenesulfonate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H14N6O3 · C7H8O3S
CAS Number:
Molecular Weight:
438.46
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>98% (TLC)

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.NC[C@H]2O[C@H]([C@H](O)[C@@H]2O)n3cnc4c(N)ncnc34

InChI

1S/C10H14N6O3.C7H8O3S/c11-1-4-6(17)7(18)10(19-4)16-3-15-5-8(12)13-2-14-9(5)16;1-6-2-4-7(5-3-6)11(8,9)10/h2-4,6-7,10,17-18H,1,11H2,(H2,12,13,14);2-5H,1H3,(H,8,9,10)/t4-,6-,7-,10-;/m1./s1

InChI key

KEAONEXDJAQJJU-MCDZGGTQSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library