Skip to Content
Merck

A7230

Alinidine

≥98% (HPLC), solid

Synonym(s):

N-(2,6-Dichlorophenyl)-4,5-dihydro-N-2-propenyl-1H-imidazol-2-amine, ST-567

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H13Cl2N3
CAS Number:
Molecular Weight:
270.16
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

storage condition

under inert gas

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Clc1cccc(Cl)c1N(CC=C)C2=NCCN2

Biochem/physiol Actions

Alinidine is an HCN Channel blocker of neuronal Ih, related cardiac If channels and ATP-sensitive Kir channels. It is an analog of clonidine; bradycardic and antiarrhythmic agent (sinus tachyarrhythmias). Alinidine affects physiological markers in conscious dogs. Alinidine in four intravenous (i.v.) injections of 0.5, 0.5, 1, and 2 mg/kg, decreased sinus rate (< or = 43%) and ventricular rate (< or = 44%), but increased atrial rate (< or = 31%). It lengthened CSRT (< or = 71%) at the two highest doses and increased AERP (< or = 33%) and decreased WP (< or = 33%) at all doses. Alinidine did not modify mean blood pressure at any dose in either group. These results indicate that alinidine exhibits electrophysiologic effects in conscious dogs that reflect the marked antiarrhythmic potential of this agent, apart from its assumed antiischemic properties.
Alinidine is an HCN channel blocker; Blocker of neuronal Ih, related cardiac If channels and ATP-sensitive Kir channels. Analog of clonidine; bradycardic and antiarrhythmic agent (sinus tachyarrhythmias).

Disclaimer

Air-sensitive


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mirko Baruscotti et al.
Pharmacology & therapeutics, 107(1), 59-79 (2005-06-21)
First described over a quarter of a century ago, the cardiac pacemaker "funny" (I(f)) current has been extensively characterized since, and its role in cardiac pacemaking has been thoroughly demonstrated. A similar current, termed I(h), was later described in different
R E BoSmith et al.
British journal of pharmacology, 110(1), 343-349 (1993-09-01)
1. ZENECA ZD7288 (4-(N-ethyl-N-phenylamino)-1,2-dimethyl-6-(methylamino) pyridinium chloride) is a sinoatrial node (SAN) modulating agent which produces a selective slowing of the heart rate. Its effects have been studied in single, freshly dissociated guinea-pig SAN cells, by standard patch clamp procedures. 2.
N J Lodge et al.
Naunyn-Schmiedeberg's archives of pharmacology, 354(4), 444-451 (1996-10-01)
The goal of the present study was to further characterize the effects of the novel cardioprotective agent BMS-180448 on potassium fluxes in cardiac and vascular smooth muscle. Exposure of voltage-clamped guinea pig ventricular myocytes to BMS-180448 (300 microM) produced an