Skip to Content
Merck

Skip To

A7169

Avidin–TRITC Conjugate

buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

TRITC conjugate

form

buffered aqueous solution

technique(s)

indirect immunofluorescence: 1:100

storage temp.

−20°C

Physical form

Solution in 0.01 M phosphate buffered saline, containing 10% (v/v) 0.5 M carbonate buffer, pH 9.5 and 15 mM sodium azide.


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library