Skip to Content
Merck
All Photos(1)

Key Documents

A7061

Sigma-Aldrich

5β-Androstane-3α,17α-diol-11-one-17β-carboxylic acid 3-(β-D-glucuronide) disodium salt

Synonym(s):

Etiocholane-3α,17α-diol-11-one-17β-carboxylic acid 3-(β-D-glucuronide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H36O11Na2
CAS Number:
Molecular Weight:
570.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

shipped in

ambient

storage temp.

room temp

SMILES string

[Na].CC12CCC(CC1CCC3C4CCC(O)(C(O)=O)C4(C)CC(=O)C23)OC5OC(C(O)C(O)C5O)C(O)=O

InChI

1S/C26H38O11.Na.H/c1-24-7-5-12(36-22-19(30)17(28)18(29)20(37-22)21(31)32)9-11(24)3-4-13-14-6-8-26(35,23(33)34)25(14,2)10-15(27)16(13)24;;/h11-14,16-20,22,28-30,35H,3-10H2,1-2H3,(H,31,32)(H,33,34);;

InChI key

KNMXCTWKUXBAPR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service