Skip to Content
Merck

A6583

Ala-Pro-Arg-Thr-Pro-Gly-Gly-Arg-Arg

synthetic, lyophilized powder

Synonym(s):

S5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)O)C(=O)N2CCC[C@H]2C(=O)NCC(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C39H70N18O11/c1-20(40)34(65)56-16-7-12-26(56)33(64)53-23(9-4-14-48-38(43)44)31(62)55-29(21(2)58)35(66)57-17-6-11-25(57)32(63)51-18-27(59)50-19-28(60)52-22(8-3-13-47-37(41)42)30(61)54-24(36(67)68)10-5-15-49-39(45)46/h20-26,29,58H,3-19,40H2,1-2H3,(H,50,59)(H,51,63)(H,52,60)(H,53,64)(H,54,61)(H,55,62)(H,67,68)(H4,41,42,47)(H4,43,44,48)(H4,45,46,49)/t20-,21+,22-,23-,24-,25-,26-,29-/m0/s1

InChI key

UAPJQIXAXHXPGL-XACXWONYSA-N

Gene Information

human ... EDN1(1906)

Biochem/physiol Actions

Substrate for meiosis-activated myelin basic protein kinase (p44mpk)


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



I Clark-Lewis et al.
The Journal of biological chemistry, 266(23), 15180-15184 (1991-08-15)
Synthetic peptides have been used to define the consensus amino acid sequence for substrate recognition by the meiosis-activated myelin basic protein (MBP) kinase (p44mpk), which was purified from maturing sea star oocytes. This protein kinase shares many properties with the