Skip to Content
Merck

A5806

9-Amino-6-chloro-2-methoxyacridine

suitable for fluorescence, ≥95%

Synonym(s):

ACMA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H11ClN2O
CAS Number:
Molecular Weight:
258.70
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95%

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

COc1ccc2nc3cc(Cl)ccc3c(N)c2c1

InChI

1S/C14H11ClN2O/c1-18-9-3-5-12-11(7-9)14(16)10-4-2-8(15)6-13(10)17-12/h2-7H,1H3,(H2,16,17)

InChI key

IHHSSHCBRVYGJX-UHFFFAOYSA-N



pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library