Skip to Content
Merck

A4895

Acid Black 48

practical grade

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Colour Index Number:
65005
EC Number:
215-518-4
UNSPSC Code:
12171500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

practical grade





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library