Skip to Content
Merck

A3952

Acetonitrile solution

5% in H2O, liquid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
4124158
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

concentration

5% in H2O

density

0.782 g/mL at 25 °C, 0.808 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Acetonitrile solution




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their
B Girit et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 71(12), 1859-1868 (2015-06-13)
Bisphenol A (BPA) is an industrial pollutant considered as one of the major endocrine-disrupting chemicals found in natural waters. In the present study, the use of a commercial, air-stable, zero-valent iron (ZVI) powder, consisting of Fe0 surface stabilized nanoparticles was
Takashi Fujii et al.
Nature protocols, 10(9), 1445-1456 (2015-08-28)
Live single-cell mass spectrometry (live MS) provides a mass spectrum that shows thousands of metabolite peaks from a single live plant cell within minutes. By using an optical microscope, a cell is chosen for analysis and a metal-coated nanospray microcapillary