Skip to Content
Merck

A3432

N2-Asn

98%

Synonym(s):

β-D-GlcNAc-(1→4)-β-D-GlcNAc-1-N-Asn, 2-Acetamido-4-O-(2-acetamido-2-deoxy-β-D-glucopyranosyl)-1-N-(β-L-aspartyl)-2-deoxy-β-D-glucopyranosylamine, N,N′-Diacetyl-1-(N-β-L-aspartyl)-β-chitobiose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H34N4O13
CAS Number:
Molecular Weight:
538.50
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

98%

storage temp.

−20°C

SMILES string

CC(=O)NC1C(O)C(OC2OC(CO)C(O)C(O)C2NC(C)=O)C(CO)OC1NC(=O)CC(N)C(O)=O





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library