Skip to Content
Merck

A3097

[Pro18, Asp21]-Amyloid β Protein Fragment 17-21 trifluoroacetate salt

≥98% (HPLC), solid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H43N5O8
CAS Number:
Molecular Weight:
637.72
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](N)C(=O)N1CCC[C@H]1C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CC(O)=O)C(O)=O

Gene Information

human ... APP(351)

Biochem/physiol Actions

Inhibits amyloid formation in vitro and amyloid neurotoxicity in cell culture, reduces in vivo cerebral amyloid deposition and blocks amyloid fibrial formation in rat brain.
Inhibits amyloid formation in vitro, prevents amyloid neurotoxicity in cell culture, reduces in vivo cerebral amyloid deposition and completely blocks amyloid fibrial formation in rat brain.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library