Skip to Content

A3028

Apocholic acid

Synonym(s):

3α,12α-Dihydroxy-5β,8(14)-cholen-24-oic acid, 3α,12α-Dihydroxy-5β-chol-8(14)en-24-oic acid, 5β,8(14)-Cholen-24-oic acid-3α,12α-diol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

175-176 °C (lit.)

SMILES string

C[C@H](CCC(O)=O)[C@H]1CCC2=C3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library