A2960
D-Ala-β-(2-naphthyl)-D-Ala-Trp-D-Phe-Lys amide
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C42H50N8O5
Molecular Weight:
746.90
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
C[C@@H](N)C(=O)N[C@H](Cc1ccc2ccccc2c1)C(=O)N[C@@H](Cc3c[nH]c4ccccc34)C(=O)N[C@H](Cc5ccccc5)C(=O)N[C@@H](CCCCN)C(N)=O
Biochem/physiol Actions
Growth hormone-releasing peptide studied in rats
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H Sawada et al.
Regulatory peptides, 53(3), 195-201 (1994-10-21)
The effect of a newly developed growth hormone (GH)-releasing hexapeptide (KP-102) on GH secretion was studied in urethan-anesthetized adult male rats. Although KP-102 alone exerted a small influence on GH secretion, it produced a large plasma GH response in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service