Skip to Content

A1675

Arg-Phe-Asp-Ser

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H33N7O8
CAS Number:
Molecular Weight:
523.54
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

NC(CCCNC(N)=N)C(=O)NC(Cc1ccccc1)C(=O)NC(CC(O)=O)C(=O)NC(CO)C(O)=O

InChI

1S/C22H33N7O8/c23-13(7-4-8-26-22(24)25)18(33)27-14(9-12-5-2-1-3-6-12)19(34)28-15(10-17(31)32)20(35)29-16(11-30)21(36)37/h1-3,5-6,13-16,30H,4,7-11,23H2,(H,27,33)(H,28,34)(H,29,35)(H,31,32)(H,36,37)(H4,24,25,26)

InChI key

SAGYNNXCQOFQHW-UHFFFAOYSA-N

Biochem/physiol Actions

Blocks integrin-binding and nonbinding of fibronectin fragments

Other Notes

Peptide analog of Arg-Gly-Asp-Ser


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library