Skip to Content
Merck

A131

Alprenolol hydrochloride

Synonym(s):

1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H23NO2 · HCl
CAS Number:
Molecular Weight:
285.81
EC Number:
237-244-4
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CC(C)NCC(O)COc1ccccc1CC=C

InChI

1S/C15H23NO2.ClH/c1-4-7-13-8-5-6-9-15(13)18-11-14(17)10-16-12(2)3;/h4-6,8-9,12,14,16-17H,1,7,10-11H2,2-3H3;1H

InChI key

RRCPAXJDDNWJBI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library