Skip to Content

A1300

L-(+)-2-Amino-5-phosphonopentanoic acid

~95%

Synonym(s):

L-AP-5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H12NO5P
CAS Number:
Molecular Weight:
197.13
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95%

storage temp.

2-8°C

SMILES string

N[C@@H](CCCP(O)(O)=O)C(O)=O

Other Notes

Inactive enantiomer of 2-amino-5-phosphonopentanoic acid.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library