Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H20N2O7
CAS Number:
Molecular Weight:
328.32
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
5298835
MDL number:
assay
≥97% (elemental analysis)
mp
148-151 °C (lit.)
SMILES string
Nc1cc2OCCOCCOCCOCCOc2cc1[N+]([O-])=O
InChI
1S/C14H20N2O7/c15-11-9-13-14(10-12(11)16(17)18)23-8-6-21-4-2-19-1-3-20-5-7-22-13/h9-10H,1-8,15H2
InChI key
PSFJQUGCUJJHIS-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.