Skip to Content
Merck

A0828

Sialyllactose sodium salt

from human milk, ≥97% (HPAE/PAD)

Synonym(s):

α-NeuNAc-(2→3)- and -(2→6)-β-D-Gal-(1→4)-D-Glc, N-Acetylneuraminyl-lactose, Neuramin-lactose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H38NO19Na
CAS Number:
Molecular Weight:
655.53
PubChem Substance ID:
UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human milk

assay

≥97% (HPAE/PAD)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H][C@]1(O[C@](C[C@H](O)[C@H]1NC(C)=O)(OC[C@H]2OC(O)[C@H](O)[C@@H](O)[C@@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)C([O-])=O)[C@H](O)[C@H](O)CO.[H][C@]4(O[C@](C[C@H](O)[C@H]4NC(C)=O)(O[C@@H]5[C@@H](O)C(O)O[C@H](CO)[C@H]5O[C@@H]6O[C@H](CO)[C@H](O)[C@H](O)[C@H]6O)C([O-])=O)[C@H](O)[C@H](O)CO

InChI

1S/2C23H39NO19.2Na/c1-6(27)24-11-7(28)2-23(22(37)38,43-19(11)12(30)8(29)3-25)39-5-10-18(15(33)16(34)20(36)40-10)42-21-17(35)14(32)13(31)9(4-26)41-21;1-6(28)24-11-7(29)2-23(22(37)38,42-18(11)12(31)8(30)3-25)43-19-16(35)20(36)39-10(5-27)17(19)41-21-15(34)14(33)13(32)9(4-26)40-21;;/h7-21,25-26,28-36H,2-5H2,1H3,(H,24,27)(H,37,38);7-21,25-27,29-36H,2-5H2,1H3,(H,24,28)(H,37,38);;/q;;2*+1/p-2/t7-,8+,9+,10+,11+,12+,13-,14-,15+,16+,17+,18+,19+,20?,21-,23-;7-,8+,9+,10+,11+,12+,13-,14-,15+,16+,17+,18+,19+,20?,21-,23+;;/m00../s1

InChI key

CFDGOUWSZLIXEU-IAQYHCRJSA-L

Application

3′-Sialyllactose is a compound wherein the acetylneuraminyl (NANA) unit is connected to the galactosyl unit of lactose at the 3 position. In 6′-sialylactose, this connection is at the 6 position. 3′-Sialyllactose and 6′-Sialyllactose are used to differentiate and characterize binding domains of viruses, such as influenza and rhinitis viruses, that recognize NANA capped cell surface receptors. 3′-Sialyllactose and 6′-sialylactose are used as reference compounds in analytical methods developed to measure their presence in materials such as milk and colostrums.

Other Notes

Mixture of (2→6′) and (2→3′) isomers.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library